Search
Search

CPB2| Gene Symbol: CPB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278541.1 | NP_001265470.1 | 8, 9 | 951 | ||
| NM_001872.4 | NP_001863.3 | 9, 10 | 1062 | |||
| XM_017020393.1 | XP_016875882.1 | 9, 10 | 1005 | |||
| GenBank mRNA | AB011969.1 | - | 8, 9 | 903 | ||
| AK290829.1 | - | 9, 10 | 1010 | |||
| BC007057.1 | - | 9, 10 | 1019 | 3934520 | ||
| BT006936.1 | - | 9, 10 | 995 | |||
| M75106.1 | - | 9, 10 | 1014 | |||