Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other EGF SIRNA Assays ›
          Gene Symbol:
          EGF
          Gene Name:
          epidermal growth factor
          Gene Aliases:
          HOMG4, URG
          Chromosome Location:
          Chr.4: 109912883 - 110013079 on Build GRCh38
          UniGene ID:
          Hs.419815
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146220

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: EGF
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001178130.2 NP_001171601.1 9, 10 1898
          NM_001178131.2 NP_001171602.1 8, 9 1772
          NM_001963.5 NP_001954.2 9, 10 1898
          XM_005262796.2 XP_005262853.1 9, 10 1899
          XM_011531707.1 XP_011530009.1 10, 11 1548
          XM_017007845.1 XP_016863334.1 9, 10 1923
          XM_017007846.1 XP_016863335.1 9, 10 1923
          XM_017007847.1 XP_016863336.1 9, 10 1923
          XM_017007848.1 XP_016863337.1 8, 9 1797
          XM_017007849.1 XP_016863338.1 9, 10 1923
          XM_017007850.1 XP_016863339.1 9, 10 1923
          XM_017007851.1 XP_016863340.1 9, 10 1923
          XM_017007852.1 XP_016863341.1 8, 9 1773
          XM_017007853.1 XP_016863342.1 9, 10 1923
          XM_017007854.1 XP_016863343.1 9, 10 1923
          XM_017007855.1 XP_016863344.1 9, 10 1923
          GenBank mRNA AK299306.1 - 8, 9 1774
          BC093731.1 - 9, 10 1541 7939576
          BC113461.1 - 9, 10 1541 8322513
          BC143357.1 - 9, 10 1541
          X04571.1 - 9, 10 1882

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          activation of MAPKK activity
          activation of MAPK activity
          angiogenesis
          platelet degranulation
          DNA replication
          signal transduction
          activation of transmembrane receptor protein tyrosine kinase activity
          epidermal growth factor receptor signaling pathway
          STAT protein import into nucleus
          positive regulation of cell proliferation
          positive regulation of peptidyl-threonine phosphorylation
          regulation of phosphatidylinositol 3-kinase signaling
          peptidyl-tyrosine phosphorylation
          positive regulation of cerebellar granule cell precursor proliferation
          positive regulation of catenin import into nucleus
          ERBB2 signaling pathway
          negative regulation of epidermal growth factor receptor signaling pathway
          positive regulation of phosphorylation
          positive regulation of DNA binding
          positive regulation of MAP kinase activity
          positive regulation of GTPase activity
          Wnt signaling pathway involved in dorsal/ventral axis specification
          positive regulation of epidermal growth factor-activated receptor activity
          positive regulation of mitotic nuclear division
          positive regulation of transcription, DNA-templated
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          branching morphogenesis of an epithelial tube
          negative regulation of secretion
          canonical Wnt signaling pathway
          mammary gland alveolus development
          ERK1 and ERK2 cascade
          regulation of calcium ion import
          negative regulation of cholesterol efflux
          positive regulation of hyaluronan biosynthetic process
          regulation of protein localization to cell surface
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          regulation of cell motility
          protein tyrosine kinase activity
          Ras guanyl-nucleotide exchange factor activity
          epidermal growth factor receptor binding
          calcium ion binding
          protein binding
          growth factor activity
          Wnt-protein binding
          transmembrane receptor protein tyrosine kinase activator activity
          Wnt-activated receptor activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline