Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other FGF2 SIRNA Assays ›
Gene Symbol:
FGF2
Gene Name:
fibroblast growth factor 2
Gene Aliases:
BFGF, FGF-2, FGFB, HBGF-2
Chromosome Location:
Chr.4: 122826708 - 122898235 on Build GRCh38
UniGene ID:
Hs.284244
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
146273
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: FGF2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_002006.4
NP_001997.5
3
4620
GenBank mRNA
BP292299.1
-
1
136
J04513.1
-
3
4617
Back To Top
More Information
Panther Classification:
Molecular Function -
growth factor
intercellular signal molecule
Gene Ontology Categories:
Function(s)
Process(es)
MAPK cascade
activation of MAPK activity
branching involved in ureteric bud morphogenesis
positive regulation of endothelial cell proliferation
cell migration involved in sprouting angiogenesis
phosphatidylinositol biosynthetic process
chemotaxis
signal transduction
Ras protein signal transduction
nervous system development
positive regulation of cell proliferation
fibroblast growth factor receptor signaling pathway
organ morphogenesis
negative regulation of fibroblast migration
positive regulation of phospholipase C activity
regulation of phosphatidylinositol 3-kinase signaling
growth factor dependent regulation of skeletal muscle satellite cell proliferation
peptidyl-tyrosine phosphorylation
extracellular matrix organization
hyaluronan catabolic process
inositol phosphate biosynthetic process
somatic stem cell population maintenance
phosphatidylinositol-3-phosphate biosynthetic process
wound healing
positive regulation of cell fate specification
positive regulation of MAP kinase activity
positive regulation of blood vessel endothelial cell migration
negative regulation of blood vessel endothelial cell migration
positive regulation of GTPase activity
positive regulation of phosphatidylinositol 3-kinase activity
regulation of angiogenesis
positive regulation of angiogenesis
positive regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
phosphatidylinositol phosphorylation
phosphatidylinositol-mediated signaling
embryonic morphogenesis
positive chemotaxis
release of sequestered calcium ion into cytosol
positive regulation of cell division
positive regulation of cardiac muscle cell proliferation
negative regulation of cell death
chondroblast differentiation
negative regulation of wound healing
positive regulation of ERK1 and ERK2 cascade
positive regulation of sprouting angiogenesis
regulation of endothelial cell chemotaxis to fibroblast growth factor
positive regulation of endothelial cell chemotaxis to fibroblast growth factor
protein tyrosine kinase activity
Ras guanyl-nucleotide exchange factor activity
fibroblast growth factor receptor binding
cytokine activity
protein binding
growth factor activity
heparin binding
1-phosphatidylinositol-3-kinase activity
ligand-dependent nuclear receptor transcription coactivator activity
chemoattractant activity
phosphatidylinositol-4,5-bisphosphate 3-kinase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0