Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CD24 SIRNA Assays ›
          Gene Symbol:
          CD24
          Gene Name:
          CD24 molecule
          Gene Aliases:
          CD24A
          Chromosome Location:
          Chr.6: 106969831 - 106976855 on Build GRCh38
          UniGene ID:
          Hs.644105
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146712

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CD24
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001291737.1 NP_001278666.1 3 2044
          NM_001291738.1 NP_001278667.1 3 1899
          NM_001291739.1 NP_001278668.1 2 1978
          NM_013230.3 NP_037362.1 3 2117
          NR_117089.1 2 1867
          NR_117090.1 2 1940
          GenBank mRNA AK026603.1 - 2 1766
          AK125531.1 - 3 1978
          BC064619.1 - 2 1742 5591617
          BG260536.1 - 1 244
          D87667.1 - 1 980
          DQ530230.1 - 1 499
          DQ530231.1 - 1 529
          DQ530232.1 - 1 529
          DQ530233.1 - 1 484
          DQ530234.1 - 1 529
          L33930.1 - 2 1729
          M58664.1 - 2 1728

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          response to hypoxia
          cell activation
          regulation of cytokine-mediated signaling pathway
          response to molecule of bacterial origin
          immune response-regulating cell surface receptor signaling pathway
          positive regulation of cytosolic calcium ion concentration
          Wnt signaling pathway
          single organismal cell-cell adhesion
          cell migration
          regulation of epithelial cell differentiation
          T cell costimulation
          B cell receptor transport into membrane raft
          chemokine receptor transport out of membrane raft
          negative regulation of transforming growth factor beta3 production
          positive regulation of activated T cell proliferation
          regulation of phosphorylation
          cholesterol homeostasis
          positive regulation of MAP kinase activity
          regulation of MAPK cascade
          response to estrogen
          respiratory burst
          positive regulation of protein tyrosine kinase activity
          glomerular visceral epithelial cell differentiation
          glomerular parietal epithelial cell differentiation
          intrinsic apoptotic signaling pathway
          positive regulation of nephron tubule epithelial cell differentiation
          signal transducer activity
          protein binding
          protein kinase binding
          protein tyrosine kinase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline