Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CBLB SIRNA Assays ›
          Gene Symbol:
          CBLB
          Gene Name:
          Cbl proto-oncogene B
          Gene Aliases:
          Cbl-b, Nbla00127, RNF56
          Chromosome Location:
          Chr.3: 105655461 - 105869552 on Build GRCh38
          UniGene ID:
          Hs.430589
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146838

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CBLB
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001321786.1 NP_001308715.1 3 484
          NM_001321788.1 NP_001308717.1 3 403
          NM_001321789.1 NP_001308718.1 3 422
          NM_001321790.1 NP_001308719.1 3 357
          NM_001321791.1 NP_001308720.1 3 403
          NM_001321793.1 NP_001308722.1 3 579
          NM_001321794.1 NP_001308723.1 3 403
          NM_001321795.1 NP_001308724.1 3 579
          NM_001321796.1 NP_001308725.1 3 403
          NM_001321797.1 NP_001308726.1 3 403
          NM_001321798.1 NP_001308727.1 3 579
          NM_001321799.1 NP_001308728.1 3 579
          NM_001321806.1 NP_001308735.1 3 403
          NM_001321807.1 NP_001308736.1 3 579
          NM_001321808.1 NP_001308737.1 3 403
          NM_001321811.1 NP_001308740.1 3 579
          NM_001321813.1 NP_001308742.1 2 271
          NM_001321816.1 NP_001308745.1 3 579
          NM_001321820.1 NP_001308749.1 3 403
          NM_001321822.1 NP_001308751.1 3 579
          NM_170662.4 NP_733762.2 3 579
          NR_135806.1 3 579
          NR_135807.1 3 579
          NR_135808.1 3 579
          NR_135809.1 3 579
          NR_135810.1 3 357
          NR_135811.1 3 403
          NR_135812.1 3 299
          XM_011513257.1 XP_011511559.1 3 399
          XM_011513259.2 XP_011511561.2 3 524
          XM_017007395.1 XP_016862884.1 3 351
          XM_017007396.1 XP_016862885.1 3 522
          XM_017007397.1 XP_016862886.1 3 523
          XM_017007398.1 XP_016862887.1 3 349
          XM_017007399.1 XP_016862888.1 3 574
          XM_017007400.1 XP_016862889.1 3 523
          GenBank mRNA AK292792.1 - 3 520
          AK302533.1 - 3 357
          AK302892.1 - 3 335
          AK316276.1 - 3 337
          BC032851.2 - 3 520 5269036
          DQ349203.1 - 2 257
          DQ349204.1 - 2 257
          U26710.1 - 3 579
          U26711.1 - 3 579
          U26712.1 - 3 579

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ligase metabolite interconversion enzyme

          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          NLS-bearing protein import into nucleus
          signal transduction
          cell surface receptor signaling pathway
          response to gravity
          response to hormone
          regulation of signaling
          phosphotyrosine binding
          ubiquitin-protein transferase activity
          signal transducer activity
          calcium ion binding
          protein binding
          zinc ion binding
          ligase activity
          protein kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0