Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RUNX1T1 SIRNA Assays ›
          Gene Symbol:
          RUNX1T1
          Gene Name:
          RUNX1 translocation partner 1
          Gene Aliases:
          AML1-MTG8, AML1T1, CBFA2T1, CDR, ETO, MTG8, ZMYND2
          Chromosome Location:
          Chr.8: 91954967 - 92103365 on Build GRCh38
          UniGene ID:
          Hs.368431
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 146846

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: RUNX1T1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001198625.1 NP_001185554.1 10 1478
          NM_001198626.1 NP_001185555.1 8 1336
          NM_001198627.1 NP_001185556.1 9 1396
          NM_001198628.1 NP_001185557.1 7 993
          NM_001198629.1 NP_001185558.1 7 1119
          NM_001198630.1 NP_001185559.1 8 1179
          NM_001198631.1 NP_001185560.1 7 1192
          NM_001198632.1 NP_001185561.1 7 1110
          NM_001198633.1 NP_001185562.1 6 1081
          NM_001198634.1 NP_001185563.1 6 1019
          NM_001198679.1 NP_001185608.1 7 1163
          NM_004349.3 NP_004340.1 6 1129
          NM_175634.2 NP_783552.1 7 1254
          NM_175635.2 NP_783553.1 6 946
          NM_175636.2 NP_783554.1 5 1028
          XM_006716676.3 XP_006716739.1 8 1063
          XM_011517351.2 XP_011515653.1 8 1108
          XM_011517352.2 XP_011515654.1 8 1348
          XM_011517353.2 XP_011515655.1 7 1056
          XM_017013930.1 XP_016869419.1 7 1364
          XM_017013931.1 XP_016869420.1 6 886
          XM_017013932.1 XP_016869421.1 8 1251
          XM_017013933.1 XP_016869422.1 9 1315
          XM_017013934.1 XP_016869423.1 9 1423
          XM_017013935.1 XP_016869424.1 8 1228
          XM_017013936.1 XP_016869425.1 7 1322
          XM_017013937.1 XP_016869426.1 8 1148
          XM_017013938.1 XP_016869427.1 7 1012
          XM_017013939.1 XP_016869428.1 2 779
          XM_017013940.1 XP_016869429.1 3 2008
          XM_017013941.1 XP_016869430.1 2 1798
          GenBank mRNA AK297616.1 - 6 843
          AK312592.1 - 10 1240
          BC005850.1 - 6 908 2961112
          BC067078.1 - 6 899 2964656
          BC139783.1 - 2 1721
          BT009871.1 - 5 688
          CR456792.1 - 5 718
          D14289.1 - 5 985
          D14821.1 - 6 883
          D43638.1 - 6 1129
          KF946506.1 - 4 279
          KF946507.1 - 6 747
          KF946508.1 - 6 747
          KF946510.1 - 6 747
          KF946511.1 - 6 733
          KF946512.1 - 7 2037
          KF946513.1 - 6 2417
          KF946514.1 - 6 733
          KF946516.1 - 5 973
          KF946517.1 - 5 973
          KF946518.1 - 5 973
          KF946520.1 - 2 395
          KF946521.1 - 2 395
          KF946522.1 - 2 395
          KF946523.1 - 2 395
          X79990.1 - 5 711

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001108657, XM_006237903, XM_008763549, XM_006237904, XM_008763548, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001111026, NM_009822, NM_001111027, AK032132, BC139201, and others.

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          generation of precursor metabolites and energy
          transcription, DNA-templated
          negative regulation of fat cell differentiation
          negative regulation of transcription, DNA-templated
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          protein binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0