Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNA1C SIRNA Assays ›
Gene Symbol:
CACNA1C
Gene Name:
calcium voltage-gated channel subunit alpha1 C
Gene Aliases:
CACH2, CACN2, CACNL1A1, CCHL1A1, CaV1.2, LQT8, TS
Chromosome Location:
Chr.12: 1969677 - 2697949 on Build GRCh38
UniGene ID:
Hs.118262
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
147003
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNA1C
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000719.6
NP_000710.5
14
2381
NM_001129827.1
NP_001123299.1
14
2381
NM_001129829.1
NP_001123301.1
14
2381
NM_001129830.2
NP_001123302.2
14
2346
NM_001129831.1
NP_001123303.1
14
2381
NM_001129832.1
NP_001123304.1
14
2381
NM_001129833.1
NP_001123305.1
14
2381
NM_001129834.1
NP_001123306.1
14
2381
NM_001129835.1
NP_001123307.1
14
2381
NM_001129836.1
NP_001123308.1
14
2381
NM_001129837.1
NP_001123309.1
14
2381
NM_001129838.1
NP_001123310.1
14
2381
NM_001129839.1
NP_001123311.1
14
2381
NM_001129840.1
NP_001123312.1
14
2381
NM_001129841.1
NP_001123313.1
14
2381
NM_001129842.1
NP_001123314.1
14
2381
NM_001129843.1
NP_001123315.1
14
2381
NM_001129844.1
NP_001123316.1
14
2372
NM_001129846.1
NP_001123318.1
14
2381
NM_001167623.1
NP_001161095.1
14
2381
NM_001167624.2
NP_001161096.2
14
2346
NM_001167625.1
NP_001161097.1
14
2381
NM_199460.3
NP_955630.3
14
2346
XM_006719017.2
XP_006719080.1
14
2435
XM_011521020.2
XP_011519322.1
15
2510
XM_011521023.2
XP_011519325.1
15
2695
XM_017019926.1
XP_016875415.1
17
3817
XM_017019927.1
XP_016875416.1
17
3817
XM_017019928.1
XP_016875417.1
17
3817
XM_017019929.1
XP_016875418.1
17
3817
XM_017019930.1
XP_016875419.1
17
3817
XM_017019931.1
XP_016875420.1
17
3817
XM_017019932.1
XP_016875421.1
15
3622
XM_017019933.1
XP_016875422.1
15
3622
XM_017019934.1
XP_016875423.1
16
3697
XM_017019935.1
XP_016875424.1
16
3697
XM_017019936.1
XP_016875425.1
15
3622
XM_017019937.1
XP_016875426.1
15
3622
XM_017019938.1
XP_016875427.1
15
3622
XM_017019939.1
XP_016875428.1
15
3622
XM_017019940.1
XP_016875429.1
15
3622
XM_017019941.1
XP_016875430.1
15
3622
XM_017019942.1
XP_016875431.1
15
3622
XM_017019943.1
XP_016875432.1
15
3622
XM_017019944.1
XP_016875433.1
15
3622
XM_017019945.1
XP_016875434.1
15
3622
XM_017019946.1
XP_016875435.1
15
3622
XM_017019947.1
XP_016875436.1
15
3622
XM_017019948.1
XP_016875437.1
15
3622
XM_017019949.1
XP_016875438.1
15
3622
XM_017019950.1
XP_016875439.1
15
3622
XM_017019951.1
XP_016875440.1
15
3613
XM_017019952.1
XP_016875441.1
15
3622
XM_017019953.1
XP_016875442.1
14
2435
XM_017019954.1
XP_016875443.1
14
2435
XM_017019955.1
XP_016875444.1
16
3697
GenBank mRNA
AB209016.1
-
12
1816
AF465484.1
-
14
2333
AJ224873.1
-
14
2068
AY830711.1
-
14
2068
AY830712.1
-
14
2068
AY830713.1
-
14
2068
BC146846.1
-
14
2187
9056774
L29529.1
-
13
2000
L29534.1
-
13
1994
L29536.1
-
13
1994
Z34809.1
-
14
2068
Z34810.1
-
14
2068
Z34811.1
-
14
2068
Z34812.1
-
14
2068
Z34813.1
-
14
2068
Z34814.1
-
14
2068
Z34815.1
-
14
2068
Z34816.1
-
14
2068
Z34817.1
-
14
2068
Z34818.1
-
14
2068
Z34819.1
-
14
2068
Z34820.1
-
14
2068
Z34821.1
-
14
2068
Z34822.1
-
14
2068
Z74996.1
-
14
2068
Back To Top
More Information
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
immune system development
positive regulation of cytosolic calcium ion concentration
heart development
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
embryonic forelimb morphogenesis
calcium-mediated signaling using extracellular calcium source
camera-type eye development
regulation of insulin secretion
calcium ion transport into cytosol
cardiac conduction
calcium ion transmembrane transport via high voltage-gated calcium channel
calcium ion transmembrane transport
cardiac muscle cell action potential involved in contraction
membrane depolarization during cardiac muscle cell action potential
membrane depolarization during AV node cell action potential
cell communication by electrical coupling involved in cardiac conduction
regulation of heart rate by cardiac conduction
regulation of ventricular cardiac muscle cell action potential
membrane depolarization during atrial cardiac muscle cell action potential
voltage-gated calcium channel activity
protein binding
calmodulin binding
high voltage-gated calcium channel activity
metal ion binding
alpha-actinin binding
voltage-gated calcium channel activity involved in cardiac muscle cell action potential
voltage-gated calcium channel activity involved in AV node cell action potential
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0