Search
Search

ANPEP| Gene Symbol: ANPEP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001150.2 | NP_001141.2 | 19 | 2850 | ||
| XM_005254892.4 | XP_005254949.1 | 19 | 2858 | |||
| XM_011521473.1 | XP_011519775.1 | 19 | 3001 | |||
| GenBank mRNA | AB209918.1 | - | 19 | 2846 | ||
| AK298134.1 | - | 13 | 1320 | |||
| AK298242.1 | - | 13 | 1371 | |||
| AK298339.1 | - | 14 | 1431 | |||
| AK300946.1 | - | 15 | 1581 | |||
| BC058928.1 | - | 18 | 2651 | 6020902 | ||
| M22324.1 | - | 18 | 2678 | |||
| X13276.1 | - | 18 | 2582 | |||