Search
Search

GRK2| Gene Symbol: GRK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001619.3 | NP_001610.2 | 6, 7 | 777 | ||
| XM_011544773.1 | XP_011543075.1 | 6, 7 | 518 | |||
| GenBank mRNA | AB209588.1 | - | 5, 6 | 1133 | ||
| BC037963.1 | - | 6, 7 | 756 | 5585846 | ||
| BC090863.1 | - | 6, 7 | 745 | 6203199 | ||
| KU177896.1 | - | 6, 7 | 511 | |||
| KU177897.1 | - | 6, 7 | 511 | |||
| M80776.1 | - | 6, 7 | 526 | |||
| X61157.1 | - | 6, 7 | 618 | |||