Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CACNA1A SIRNA Assays ›
          Gene Symbol:
          CACNA1A
          Gene Name:
          calcium voltage-gated channel subunit alpha1 A
          Gene Aliases:
          APCA, BI, CACNL1A4, CAV2.1, EA2, FHM, HPCA, MHP, MHP1, SCA6
          Chromosome Location:
          Chr.19: 13206442 - 13506460 on Build GRCh38
          UniGene ID:
          Hs.501632
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 147830

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CACNA1A
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_000068.3 NP_000059.3 7 1228
          NM_001127221.1 NP_001120693.1 7 1228
          NM_001127222.1 NP_001120694.1 7 1228
          NM_001174080.1 NP_001167551.1 7 1228
          NM_023035.2 NP_075461.2 7 1228
          GenBank mRNA AB035726.1 - 7 1027
          AB035727.2 - 7 992
          AF004883.1 - 7 1228
          AF004884.1 - 7 1228
          FJ040507.1 - 7 992
          U79666.1 - 7 1209
          X99897.1 - 7 1267

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_012918, XM_008772355, XM_008772348, XM_008772359, XM_008772357, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001252060, NM_001252061, NM_007578, NM_001252059, AB066608, and others.

          Panther Classification:

          Molecular Function -

          voltage-gated ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          sulfur amino acid metabolic process
          glucose metabolic process
          positive regulation of cytosolic calcium ion concentration
          gamma-aminobutyric acid signaling pathway
          neuromuscular synaptic transmission
          synapse assembly
          adult walking behavior
          cell death
          gamma-aminobutyric acid secretion
          regulation of acetylcholine secretion, neurotransmission
          cell growth
          regulation of calcium ion-dependent exocytosis
          transmission of nerve impulse
          spinal cord motor neuron differentiation
          cerebellum maturation
          cerebellar molecular layer development
          cerebellar Purkinje cell differentiation
          vestibular nucleus development
          cellular chloride ion homeostasis
          negative regulation of hormone biosynthetic process
          regulation of ion transmembrane transport
          synaptic transmission, glutamatergic
          neurotransmitter metabolic process
          hormone metabolic process
          receptor clustering
          negative regulation of neuron apoptotic process
          behavioral response to pain
          calcium ion-regulated exocytosis of neurotransmitter
          dendrite morphogenesis
          regulation of axonogenesis
          regulation of insulin secretion
          musculoskeletal movement, spinal reflex action
          neuromuscular process controlling balance
          membrane depolarization
          rhythmic synaptic transmission
          calcium ion transmembrane transport
          membrane depolarization during action potential
          voltage-gated calcium channel activity
          calcium channel activity
          protein binding
          high voltage-gated calcium channel activity
          syntaxin binding
          metal ion binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-qm47m:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0