Search
Search

A1BG| Gene Symbol: A1BG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_130786.3 | NP_570602.2 | 2, 3 | 134 | ||
| GenBank mRNA | AB073611.1 | - | 2, 3 | 127 | ||
| AF414429.1 | - | 2, 3 | 114 | |||
| AK055885.1 | - | 2, 3 | 127 | |||
| AK289417.1 | - | 2, 3 | 98 | |||
| BC035719.1 | - | 2, 3 | 181 | 5588885 | ||
| BX325198.2 | - | 2, 3 | 134 | |||
| EU794585.1 | - | 2, 3 | 126 | |||