Search
Search

EXTL1| Gene Symbol: EXTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_004455.2 | NP_004446.2 | 7, 8 | 2306 | ||
| XM_005245779.3 | XP_005245836.1 | 6, 7 | 2229 | |||
| GenBank mRNA | BC065528.1 | - | 7, 8 | 2152 | 6150218 | |
| U67191.1 | - | 7, 8 | 2305 | |||