Search
Search

CALML4| Gene Symbol: CALML4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031733.2 | NP_001026903.2 | 1 | 814 | ||
| NM_001286694.1 | NP_001273623.1 | 1 | 814 | |||
| NM_001286695.1 | NP_001273624.1 | 1 | 814 | |||
| NM_033429.2 | NP_219501.2 | 1 | 814 | |||
| NR_104583.1 | 1 | 814 | ||||
| GenBank mRNA | AF308287.1 | - | 1 | 265 | ||
| AK092628.1 | - | 1 | 36 | |||
| AK296804.1 | - | 1 | 36 | |||
| BC009516.2 | - | 1 | 21 | |||
| BX439220.2 | - | 1 | 752 | |||
| BX640817.1 | - | 1 | 815 | |||
| DB204573.1 | - | 1 | 38 | |||