Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZFYVE27 SIRNA Assays ›
Gene Symbol:
ZFYVE27
Gene Name:
zinc finger FYVE-type containing 27
Gene Aliases:
PROTRUDIN, SPG33
Chromosome Location:
Chr.10: 97737121 - 97760907 on Build GRCh38
UniGene ID:
Hs.744075
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
149132
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZFYVE27
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001002261.3
NP_001002261.1
12
2253
NM_001002262.3
NP_001002262.1
12
2422
NM_001174119.1
NP_001167590.1
11
2322
NM_001174120.1
NP_001167591.1
10
2160
NM_001174121.1
NP_001167592.1
10
2164
NM_001174122.1
NP_001167593.1
9
2064
NM_144588.6
NP_653189.3
13
2443
XM_005269502.3
XP_005269559.1
13
2447
XM_005269503.3
XP_005269560.1
13
2451
XM_005269504.3
XP_005269561.1
13
2432
XM_005269505.4
XP_005269562.1
12
2426
XM_005269506.3
XP_005269563.1
12
2411
XM_005269508.3
XP_005269565.1
12
2376
XM_005269509.3
XP_005269566.1
12
2380
XM_005269510.3
XP_005269567.1
11
2192
XM_005269511.3
XP_005269568.1
11
2198
XM_011539254.2
XP_011537556.1
13
2422
XM_011539255.2
XP_011537557.1
13
2429
XM_017015647.1
XP_016871136.1
12
2430
XM_017015648.1
XP_016871137.1
11
2177
XM_017015649.1
XP_016871138.1
10
2171
XM_017015650.1
XP_016871139.1
13
2407
XM_017015651.1
XP_016871140.1
12
2361
XM_017015652.1
XP_016871141.1
12
2365
XM_017015653.1
XP_016871142.1
11
2340
XM_017015654.1
XP_016871143.1
11
2344
XM_017015655.1
XP_016871144.1
11
2183
GenBank mRNA
AK057481.1
-
11
1534
AK074876.1
-
12
2403
AK097945.1
-
10
2153
AL834235.1
-
12
2187
BC030621.2
-
13
2436
4818199
BX647836.1
-
11
7855
Back To Top
More Information
Panther Classification:
Molecular Function -
membrane traffic protein
membrane trafficking regulatory protein
Gene Ontology Categories:
Function(s)
Process(es)
neuron projection development
neurotrophin TRK receptor signaling pathway
protein localization to plasma membrane
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0