Search
Search

MGAT5B| Gene Symbol: MGAT5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001199172.1 | NP_001186101.1 | 2, 3 | 489 | ||
| NM_144677.2 | NP_653278.2 | 2, 3 | 489 | |||
| NM_198955.1 | NP_945193.1 | 1, 2 | 311 | |||
| XM_011524350.2 | XP_011522652.1 | 2, 3 | 567 | |||
| XM_011524352.2 | XP_011522654.1 | 2, 3 | 707 | |||
| XM_011524354.2 | XP_011522656.1 | 2, 3 | 718 | |||
| XM_017024200.1 | XP_016879689.1 | 2, 3 | 709 | |||
| GenBank mRNA | AB109185.1 | - | 2, 3 | 200 | ||
| AB114297.1 | - | 1, 2 | 311 | |||
| AB235153.1 | - | 2, 3 | 749 | |||
| BC041577.1 | - | 2, 3 | 489 | |||
| BC062354.1 | - | 2, 3 | 381 | |||