Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SERPINE1 SIRNA Assays ›
Gene Symbol:
Serpine1
Gene Name:
serine (or cysteine) peptidase inhibitor, clade E, member 1
Gene Aliases:
PAI-1, PAI1, Planh1
Chromosome Location:
Chr.5: 137061504 - 137072272 on Build GRCm38
UniGene ID:
Mm.250422
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
150178
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Serpine1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008871.2
NP_032897.2
9
1391
GenBank mRNA
BC054091.1
-
9
1388
30021925
M33960.1
-
9
1376
Back To Top
More Information
Panther Classification:
Molecular Function -
protease inhibitor
Gene Ontology Categories:
Function(s)
Process(es)
female gonad development
negative regulation of peptidase activity
regulation of receptor activity
positive regulation of gene expression
negative regulation of gene expression
negative regulation of plasminogen activation
negative regulation of endopeptidase activity
negative regulation of smooth muscle cell migration
positive regulation of blood coagulation
negative regulation of cell migration
positive regulation of interleukin-8 production
negative regulation of cell adhesion mediated by integrin
positive regulation of leukotriene production involved in inflammatory response
cellular response to drug
wound healing
regulation of cell proliferation
regulation of angiogenesis
positive regulation of angiogenesis
positive regulation of fibroblast proliferation
positive regulation of receptor-mediated endocytosis
positive regulation of inflammatory response
positive regulation of coagulation
defense response to Gram-negative bacterium
positive regulation of keratinocyte migration
negative regulation of fibrinolysis
negative regulation of vascular wound healing
cellular response to lipopolysaccharide
cellular response to interleukin-1
cellular response to transforming growth factor beta stimulus
positive regulation of monocyte chemotaxis
positive regulation of calcium ion import
negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
negative regulation of smooth muscle cell-matrix adhesion
negative regulation of endothelial cell apoptotic process
protease binding
serine-type endopeptidase inhibitor activity
receptor binding
protein binding
peptidase inhibitor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0