Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PRKCD SIRNA Assays ›
          Gene Symbol:
          Prkcd
          Gene Name:
          protein kinase C, delta
          Gene Aliases:
          AI385711, D14Ertd420e, PKC[d], PKCdelta, Pkcd
          Chromosome Location:
          Chr.14: 30595353 - 30626414 on Build GRCm38
          UniGene ID:
          Mm.2314
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 151132

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Prkcd
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_011103.3 NP_035233.1 5 674
          GenBank mRNA AB011812.1 - 4 437
          AB201453.1 - 2 276
          AB201454.1 - 2 276
          AB201455.1 - 2 276
          AB201456.1 - 2 276
          AF251036.1 - 4 437
          AK144231.1 - 5 679
          AK168768.1 - 5 669
          AK171476.1 - 5 676
          AY545076.1 - 4 437
          BC051416.1 - 5 638 6515302
          M69042.1 - 5 669
          X60304.1 - 4 450

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          non-receptor serine/threonine protein kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          protein phosphorylation
          apoptotic process
          response to oxidative stress
          cell cycle
          aspartate transport
          immunoglobulin mediated immune response
          phosphorylation
          peptidyl-threonine phosphorylation
          termination of signal transduction
          negative regulation of actin filament polymerization
          interleukin-10 production
          interleukin-12 production
          positive regulation of superoxide anion generation
          regulation of actin cytoskeleton organization
          collagen metabolic process
          negative regulation of glial cell apoptotic process
          cellular response to oxidative stress
          positive regulation of protein dephosphorylation
          intracellular signal transduction
          B cell proliferation
          neutrophil activation
          regulation of phosphorylation
          defense response to bacterium
          positive regulation of apoptotic process
          positive regulation of MAP kinase activity
          negative regulation of MAP kinase activity
          positive regulation of MAPK cascade
          positive regulation of glucose import
          negative regulation of insulin receptor signaling pathway
          protein autophosphorylation
          negative regulation of peptidyl-tyrosine phosphorylation
          negative regulation of filopodium assembly
          negative regulation of platelet aggregation
          cellular senescence
          positive regulation of phospholipid scramblase activity
          positive regulation of ceramide biosynthetic process
          positive regulation of glucosylceramide catabolic process
          positive regulation of sphingomyelin catabolic process
          positive regulation of response to DNA damage stimulus
          positive regulation of apoptotic signaling pathway
          nucleotide binding
          protein kinase activity
          protein serine/threonine kinase activity
          protein kinase C activity
          calcium-independent protein kinase C activity
          non-membrane spanning protein tyrosine kinase activity
          ATP binding
          enzyme activator activity
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          enzyme binding
          protein kinase binding
          insulin receptor substrate binding
          metal ion binding
          TIR domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0