Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTPRF SIRNA Assays ›
Gene Symbol:
Ptprf
Gene Name:
protein tyrosine phosphatase, receptor type, F
Gene Aliases:
AA591035, LAR, LARS, RPTP-LAR
Chromosome Location:
Chr.4: 118208213 - 118291400 on Build GRCm38
UniGene ID:
Mm.29855
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
151354
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ptprf
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_011213.2
NP_035343.2
6
890
GenBank mRNA
AF300943.1
-
4
553
AI385919.1
-
2
271
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of cytokine-mediated signaling pathway
protein dephosphorylation
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
signal transduction
nervous system development
negative regulation of cell proliferation
regulation of neuron projection development
dephosphorylation
cell migration
neuron projection regeneration
negative regulation of cell projection organization
peptidyl-tyrosine dephosphorylation
negative regulation of epidermal growth factor receptor signaling pathway
positive regulation of apoptotic process
positive regulation of neuron apoptotic process
negative regulation of insulin receptor signaling pathway
regulation of axon regeneration
positive regulation of dendrite morphogenesis
regulation of synapse structure or activity
negative regulation of neurotrophin TRK receptor signaling pathway
negative regulation of receptor binding
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
insulin receptor binding
heparin binding
hydrolase activity
phosphatase activity
receptor tyrosine kinase binding
protein complex binding
chondroitin sulfate proteoglycan binding
phosphate ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-55658555d6-4jhht:80/100.66.76.55:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline