Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other RXRA SIRNA Assays ›
          Gene Symbol:
          Rxra
          Gene Name:
          retinoid X receptor alpha
          Gene Aliases:
          9530071D11Rik, Nr2b1, RXRalpha1
          Chromosome Location:
          Chr.2: 27676324 - 27763319 on Build GRCm38
          UniGene ID:
          Mm.24624
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 151540

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Rxra
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001290481.1 NP_001277410.1 6 930
          NM_001290482.1 NP_001277411.1 6 967
          NM_011305.3 NP_035435.1 6 1109
          GenBank mRNA AK031034.1 - 6 978
          AK144675.1 - 6 1111
          BC138800.1 - 7 960 8861822
          BC138802.1 - 7 960 8861824
          M84817.1 - 7 972
          U77683.1 - 6 940
          U77684.1 - 6 970
          X66223.1 - 6 1054

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_012805, AM295012, AM295013, L06482, AM392401.

          Panther Classification:

          Molecular Function -

          C4 zinc finger nuclear receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          in utero embryonic development
          placenta development
          maternal placenta development
          heart morphogenesis
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          regulation of transcription from RNA polymerase II promoter
          transcription from RNA polymerase II promoter
          heart development
          embryo implantation
          negative regulation of cell proliferation
          negative regulation of gene expression
          modulation by virus of host morphology or physiology
          regulation of myelination
          response to retinoic acid
          peroxisome proliferator activated receptor signaling pathway
          camera-type eye development
          positive regulation of apoptotic process
          steroid hormone mediated signaling pathway
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          positive regulation of translational initiation by iron
          retinoic acid receptor signaling pathway
          protein homotetramerization
          response to glucocorticoid
          cardiac muscle cell differentiation
          ventricular cardiac muscle tissue morphogenesis
          ventricular cardiac muscle cell differentiation
          cardiac muscle cell proliferation
          mesenchyme development
          secretory columnal luminar epithelial cell differentiation involved in prostate glandular acinus development
          regulation of branching involved in prostate gland morphogenesis
          angiogenesis involved in coronary vascular morphogenesis
          visceral serous pericardium development
          positive regulation of transcription from RNA polymerase II promoter involved in cellular response to chemical stimulus
          RNA polymerase II regulatory region sequence-specific DNA binding
          DNA binding
          double-stranded DNA binding
          transcription factor activity, sequence-specific DNA binding
          steroid hormone receptor activity
          RNA polymerase II transcription factor activity, ligand-activated sequence-specific DNA binding
          9-cis retinoic acid receptor activity
          protein binding
          transcription factor binding
          zinc ion binding
          enzyme binding
          protein domain specific binding
          chromatin DNA binding
          protein complex binding
          vitamin D receptor binding
          retinoic acid receptor binding
          sequence-specific DNA binding
          transcription regulatory region DNA binding
          retinoic acid-responsive element binding
          metal ion binding
          protein heterodimerization activity
          vitamin D response element binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0