Search
Search

Suv39h1| Gene Symbol: Suv39h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290716.1 | NP_001277645.1 | 3 | 412 | ||
| NM_011514.2 | NP_035644.1 | 3 | 431 | |||
| GenBank mRNA | AF019969.1 | - | 3 | 257 | ||
| AF193861.1 | - | 3 | 253 | |||
| AF193862.1 | - | 3 | 356 | |||
| AK021174.1 | - | 3 | 413 | |||
| AK088405.1 | - | 3 | 248 | |||
| AK132238.1 | - | 3 | 293 | |||
| AK139757.1 | - | 3 | 400 | |||
| AK169389.1 | - | 3 | 239 | |||
| AK169592.1 | - | 3 | 401 | |||
| BC023860.1 | - | 3 | 308 | 5352230 | ||
| BU053459.1 | - | 3 | 431 | |||