Search
Search

Suv39h1| Gene Symbol: Suv39h1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290716.1 | NP_001277645.1 | 5 | 1180 | ||
| NM_011514.2 | NP_035644.1 | 5 | 1199 | |||
| GenBank mRNA | AF019969.1 | - | 5 | 1025 | ||
| AF193861.1 | - | 5 | 1021 | |||
| AF193862.1 | - | 5 | 1124 | |||
| AK088405.1 | - | 5 | 1016 | |||
| AK132238.1 | - | 4 | 1146 | |||
| AK169389.1 | - | 5 | 1007 | |||
| AK169592.1 | - | 5 | 1169 | |||
| BC023860.1 | - | 5 | 1076 | 5352230 | ||