Search
Search

Slc20a1| Gene Symbol: Slc20a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_015747.2 | NP_056562.1 | 11 | 2945 | ||
| GenBank mRNA | AK155506.1 | - | 11 | 2918 | ||
| BC015085.1 | - | 11 | 2845 | 4015575 | ||
| M73696.1 | - | 10 | 2863 | |||