Search
Search

Plcb1| Gene Symbol: Plcb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145830.1 | NP_001139302.1 | 3 | 559 | ||
| NM_019677.2 | NP_062651.2 | 3 | 559 | |||
| GenBank mRNA | AF498249.1 | - | 3 | 190 | ||
| AF498250.1 | - | 3 | 190 | |||
| AK040911.1 | - | 3 | 550 | |||
| AK141164.1 | - | 3 | 506 | |||
| AK141464.1 | - | 3 | 495 | |||
| AK143153.1 | - | 3 | 560 | |||
| BC058710.1 | - | 3 | 526 | 6842016 | ||
| U85712.1 | - | 3 | 193 | |||
| U85713.1 | - | 3 | 193 | |||
| U85714.1 | - | 3 | 193 | |||