Search
Search

Strn| Gene Symbol: Strn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011500.2 | NP_035630.2 | 18 | 2443 | ||
| GenBank mRNA | AJ223777.1 | - | 18 | 2370 | ||
| AK135684.1 | - | 13 | 1636 | |||
| AK136032.1 | - | 16 | 2092 | |||
| AK161520.1 | - | 1 | 126 | |||
| BC090968.1 | - | 18 | 2443 | 30646517 | ||
| BC150727.1 | - | 18 | 2411 | 9087638 | ||