Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ACSF3 SIRNA Assays ›
Gene Symbol:
Acsf3
Gene Name:
acyl-CoA synthetase family member 3
Gene Aliases:
BB101783
Chromosome Location:
Chr.8: 122775493 - 122817882 on Build GRCm38
UniGene ID:
Mm.334875
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
152602
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Acsf3
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_144932.3
NP_659181.2
10
1966
GenBank mRNA
BC021611.1
-
6
839
BC022709.1
-
9
1306
Back To Top
More Information
Panther Classification:
Molecular Function -
ligase
Gene Ontology Categories:
Function(s)
Process(es)
lipid metabolic process
fatty acid metabolic process
fatty acid biosynthetic process
metabolic process
malonate catabolic process
nucleotide binding
catalytic activity
acyl-CoA ligase activity
succinate-CoA ligase activity
ATP binding
3-oxo-2-(2'-pentenyl)cyclopentane-1-octanoic acid CoA ligase activity
fatty acid ligase activity
ligase activity
acid-thiol ligase activity
3-isopropenyl-6-oxoheptanoyl-CoA synthetase activity
2-oxo-delta3-4,5,5-trimethylcyclopentenylacetyl-CoA synthetase activity
benzoyl acetate-CoA ligase activity
2,4-dichlorobenzoate-CoA ligase activity
pivalate-CoA ligase activity
cyclopropanecarboxylate-CoA ligase activity
adipate-CoA ligase activity
citronellyl-CoA ligase activity
mentha-1,3-dione-CoA ligase activity
thiophene-2-carboxylate-CoA ligase activity
2,4,4-trimethylpentanoate-CoA ligase activity
cis-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity
trans-2-methyl-5-isopropylhexa-2,5-dienoate-CoA ligase activity
branched-chain acyl-CoA synthetase (ADP-forming) activity
aryl-CoA synthetase (ADP-forming) activity
3-hydroxypropionyl-CoA synthetase activity
perillic acid:CoA ligase (ADP-forming) activity
perillic acid:CoA ligase (AMP-forming) activity
(3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (ADP-forming) activity
(3R)-3-isopropenyl-6-oxoheptanoate:CoA ligase (AMP-forming) activity
pristanate-CoA ligase activity
malonyl-CoA synthetase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0