Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other 4930503L19RIK SIRNA Assays ›
          Gene Symbol:
          4930503L19Rik
          Gene Name:
          RIKEN cDNA 4930503L19 gene
          Gene Aliases:
          9930107F24, BB148262, Lars, Las2
          Chromosome Location:
          Chr.18: 70452210 - 70472504 on Build GRCm38
          UniGene ID:
          Mm.87470
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 153513

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: 4930503L19Rik
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_172967.2 NP_766555.1 3 658
          GenBank mRNA AK037066.1 - 3 659
          AK080780.1 - 3 689
          AK087788.1 - 3 631
          AK148410.1 - 3 682
          AK160248.1 - 3 532
          AK160714.1 - 3 628
          BC049726.1 - 170
          BC057927.1 - 2 375 5290837
          HQ438967.1 - 2 170
          HQ438968.1 - 2 170
          HQ438969.1 - 2 170
          HQ438970.1 - 2 170
          HQ438971.1 - 2 170
          HQ438972.1 - 2 170
          HQ438973.1 - 2 179
          HQ438974.1 - 2 179
          HQ438975.1 - 2 179
          HQ438976.1 - 1 129
          HQ438977.1 - 1 129
          HQ438978.1 - 1 138
          HQ438979.1 - 1 129
          HQ438980.1 - 1 129
          HQ438981.1 - 1 129
          HQ438982.1 - 1 129
          HQ438983.1 - 1 129
          HQ438984.1 - 1 138
          HQ438985.1 - 1 129
          HQ438986.1 - 1 138
          HQ438987.1 - 1 129
          HQ438988.1 - 1 129
          HQ438989.1 - 1 129
          HQ438990.1 - 1 138
          HQ438991.1 - 1 138
          HQ438992.1 - 1 129
          HQ438993.1 - 1 138
          HQ438994.1 - 1 129
          HQ438995.1 - 1 138
          HQ438996.1 - 1 138
          HQ438997.1 - 1 138
          HQ438998.1 - 2 179
          HQ438999.1 - 1 129
          HQ439000.1 - 1 129
          HQ439001.1 - 1 138
          HQ439002.1 - 1 129
          HQ439003.1 - 1 129
          HQ439004.1 - 1 138
          HQ439005.1 - 1 129
          HQ439006.1 - 1 129
          HQ439007.1 - 1 138
          HQ439008.1 - 1 138
          HQ439009.1 - 1 129
          HQ439010.1 - 1 138
          HQ439011.1 - 1 138
          HQ439012.1 - 1 129

          Back To Top

          More Information



          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0