Search
Search

Zdhhc19| Gene Symbol: Zdhhc19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199309.2 | NP_955013.1 | 7 | 841 | ||
| GenBank mRNA | AK132780.1 | - | 7 | 858 | ||
| AK133398.1 | - | 7 | 842 | |||
| BC049761.1 | - | 7 | 841 | 6773873 | ||
| BC141059.1 | - | 6 | 750 | 9055671 | ||