Search
Search

Mep1a| Gene Symbol: Mep1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008585.2 | NP_032611.2 | 8 | 731 | ||
| GenBank mRNA | AK093914.1 | - | 8 | 692 | ||
| AK143906.1 | - | 8 | 689 | |||
| AK164734.1 | - | 8 | 686 | |||
| BC015258.1 | - | 8 | 731 | 4241252 | ||
| M74897.1 | - | 8 | 730 | |||