Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MET SIRNA Assays ›
          Gene Symbol:
          Met
          Gene Name:
          met proto-oncogene
          Gene Aliases:
          AI838057, HGF, HGFR, Par4, c-Met
          Chromosome Location:
          Chr.6: 17463349 - 17573980 on Build GRCm38
          UniGene ID:
          Mm.86844
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 155341

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Met
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008591.2 NP_032617.2 3 927
          GenBank mRNA AK148258.1 - 3 778
          AK170560.1 - 3 927
          BC117969.1 - 1 571
          BC117970.1 - 1 567
          Y00671.1 - 1 572

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_031517, XM_006236129, U65007, X96786.

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          activation of MAPK activity
          neuron migration
          endothelial cell morphogenesis
          liver development
          placenta development
          protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          brain development
          muscle organ development
          skeletal muscle tissue development
          positive regulation of gene expression
          positive regulation of peptidyl-threonine phosphorylation
          negative regulation of peptidyl-threonine phosphorylation
          positive regulation of glucose transport
          positive regulation of neuron projection development
          muscle cell migration
          myotube differentiation
          phosphorylation
          peptidyl-tyrosine phosphorylation
          sperm motility
          adult behavior
          positive regulation of peptidyl-serine phosphorylation
          glucose homeostasis
          positive regulation of DNA replication
          positive regulation of mitotic nuclear division
          positive regulation of transcription from RNA polymerase II promoter
          protein autophosphorylation
          hepatocyte growth factor receptor signaling pathway
          branching morphogenesis of an epithelial tube
          positive regulation of dendrite morphogenesis
          modulation of synaptic transmission
          positive chemotaxis
          myoblast proliferation
          excitatory postsynaptic potential
          regulation of branching involved in salivary gland morphogenesis by mesenchymal-epithelial signaling
          semaphorin-plexin signaling pathway
          negative regulation of hydrogen peroxide-mediated programmed cell death
          positive regulation of endothelial cell chemotaxis
          nucleotide binding
          protein kinase activity
          protein tyrosine kinase activity
          transmembrane receptor protein tyrosine kinase activity
          hepatocyte growth factor-activated receptor activity
          protein binding
          ATP binding
          beta-catenin binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein phosphatase binding
          protein complex binding
          phospholipase binding
          phosphatidylinositol 3-kinase binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0