Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBM39 SIRNA Assays ›
Gene Symbol:
RBM39
Gene Name:
RNA binding motif protein 39
Gene Aliases:
CAPER, CAPERalpha, FSAP59, HCC1, RNPC2
Chromosome Location:
Chr.20: 35703609 - 35742336 on Build GRCh38
UniGene ID:
Hs.282901
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
15556
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBM39
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242599.1
NP_001229528.1
4
576
NM_001242600.1
NP_001229529.1
4
576
NM_001323422.1
NP_001310351.1
4
576
NM_001323423.1
NP_001310352.1
6
721
NM_001323424.1
NP_001310353.1
4
576
NM_004902.3
NP_004893.1
4
576
NM_184234.2
NP_909122.1
4
576
NR_040722.1
5
649
NR_040723.1
5
648
NR_040724.1
5
649
XM_006723890.3
XP_006723953.1
4
2156
XM_006723891.3
XP_006723954.1
5
1179
XM_011529111.2
XP_011527413.1
5
2228
XM_017028138.1
XP_016883627.1
4
576
XM_017028139.1
XP_016883628.1
6
1545
XM_017028140.1
XP_016883629.1
5
649
XM_017028141.1
XP_016883630.1
4
2156
XM_017028142.1
XP_016883631.1
5
3052
XM_017028143.1
XP_016883632.1
5
2228
XM_017028144.1
XP_016883633.1
6
721
XM_017028145.1
XP_016883634.1
5
649
XM_017028146.1
XP_016883635.1
5
649
XM_017028147.1
XP_016883636.1
5
572
GenBank mRNA
AK126158.1
-
1
735
AK299165.1
-
4
476
AK299678.1
-
4
478
AK303910.1
-
4
448
AL833168.1
-
4
462
BC107886.1
-
5
358
BC131543.1
-
4
342
40085367
BC141835.1
-
4
426
40148871
BC158172.1
-
4
426
40148190
BX537770.1
-
5
568
BX640714.1
-
5
561
BX640812.1
-
5
558
CR749443.1
-
4
2243
DQ656052.2
-
4
183
L10910.1
-
4
316
L10911.1
-
3
191
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA splicing factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
RNA processing
mRNA processing
RNA splicing
nucleotide binding
protein binding
poly(A) RNA binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-q45k6:80/100.66.78.42:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0