Search
Search

Notch1| Gene Symbol: Notch1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008714.3 | NP_032740.3 | 3 | 466 | ||
| GenBank mRNA | AF508809.1 | - | 3 | 281 | ||
| AK090118.1 | - | 3 | 442 | |||
| BC138441.1 | - | 3 | 281 | 8861461 | ||
| BC138442.1 | - | 3 | 281 | 8861462 | ||