Search
Search

C21orf2| Gene Symbol: C21orf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001271440.1 | NP_001258369.1 | 7 | 1257 | ||
| NM_001271441.1 | NP_001258370.1 | 7 | 1617 | |||
| NM_001271442.1 | NP_001258371.1 | 8 | 1165 | |||
| NM_004928.2 | NP_004919.1 | 7 | 1260 | |||
| XM_006724051.3 | XP_006724114.1 | 6 | 1389 | |||
| XM_006724053.3 | XP_006724116.1 | 6 | 1066 | |||
| XM_017028470.1 | XP_016883959.1 | 7 | 1268 | |||
| XM_017028471.1 | XP_016883960.1 | 6 | 1090 | |||
| XM_017028472.1 | XP_016883961.1 | 6 | 1145 | |||
| GenBank mRNA | AB209578.1 | - | 6 | 4495 | ||
| BC031300.1 | - | 7 | 1608 | 4878257 | ||
| BC072012.1 | - | 7 | 1233 | 6645626 | ||
| U84569.1 | - | 7 | 1228 | |||
| U84570.1 | - | 8 | 1164 | |||
| Y11392.1 | - | 7 | 1120 | |||
| Z93322.1 | - | 7 | 1059 | |||