Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other INSR SIRNA Assays ›
Gene Symbol:
Insr
Gene Name:
insulin receptor
Gene Aliases:
4932439J01Rik, CD220, D630014A15Rik, IR, IR-A, IR-B
Chromosome Location:
Chr.8: 3150922 - 3279649 on Build GRCm38
UniGene ID:
Mm.268003
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
155886
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Insr
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_010568.2
NP_034698.2
3
1403
GenBank mRNA
AK052187.1
-
3
1338
J05149.1
-
3
962
KC356871.1
-
3
914
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: M10051.
This siRNA also targets the following transcripts in Rat: NM_017071, XM_006248753, XM_006248752, KF112847, KF112848, and others.
Panther Classification:
Molecular Function -
transmembrane signal receptor
Gene Ontology Categories:
Function(s)
Process(es)
activation of MAPK activity
negative regulation of protein phosphorylation
positive regulation of protein phosphorylation
heart morphogenesis
regulation of transcription, DNA-templated
protein phosphorylation
transmembrane receptor protein tyrosine kinase signaling pathway
G-protein coupled receptor signaling pathway
positive regulation of cell proliferation
insulin receptor signaling pathway
epidermis development
organ morphogenesis
regulation of hydrogen peroxide metabolic process
positive regulation of glycoprotein biosynthetic process
negative regulation of gene expression
phosphorylation
peptidyl-tyrosine phosphorylation
transformation of host cell by virus
signal transduction by protein phosphorylation
male sex determination
positive regulation of cell migration
exocrine pancreas development
activation of protein kinase activity
activation of protein kinase B activity
negative regulation of transporter activity
cellular response to insulin stimulus
response to tumor necrosis factor
intracellular signal transduction
peptidyl-tyrosine autophosphorylation
positive regulation of phosphorylation
glucose homeostasis
positive regulation of MAPK cascade
positive regulation of nitric oxide biosynthetic process
positive regulation of glycogen biosynthetic process
positive regulation of DNA replication
positive regulation of glycolytic process
positive regulation of mitotic nuclear division
regulation of embryonic development
positive regulation of glucose import
protein autophosphorylation
positive regulation of developmental growth
protein heterotetramerization
positive regulation of protein kinase B signaling
positive regulation of respiratory burst
cellular response to growth factor stimulus
nucleotide binding
protein kinase activity
protein tyrosine kinase activity
transmembrane receptor protein tyrosine kinase activity
receptor signaling protein tyrosine kinase activity
insulin-activated receptor activity
insulin-like growth factor receptor binding
protein binding
ATP binding
GTP binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
protein kinase binding
protein phosphatase binding
protein domain specific binding
lipoic acid binding
insulin-like growth factor I binding
insulin-like growth factor II binding
protein complex binding
3-phosphoinositide-dependent protein kinase binding
phosphatidylinositol 3-kinase binding
insulin binding
insulin receptor substrate binding
PTB domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0