Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NUCB2 SIRNA Assays ›
          Gene Symbol:
          NUCB2
          Gene Name:
          nucleobindin 2
          Gene Aliases:
          HEL-S-109, NEFA
          Chromosome Location:
          Chr.11: 17260340 - 17331881 on Build GRCh38
          UniGene ID:
          Hs.654599
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 15614

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: NUCB2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005013.2 NP_005004.1 5 513
          XM_017017804.1 XP_016873293.1 7 748
          XM_017017805.1 XP_016873294.1 6 664
          XM_017017806.1 XP_016873295.1 6 607
          XM_017017807.1 XP_016873296.1 6 618
          XM_017017808.1 XP_016873297.1 5 552
          XM_017017809.1 XP_016873298.1 5 549
          XM_017017810.1 XP_016873299.1 7 1019
          XM_017017811.1 XP_016873300.1 7 928
          XM_017017812.1 XP_016873301.1 5 513
          XM_017017813.1 XP_016873302.1 6 612
          XM_017017814.1 XP_016873303.1 6 605
          XM_017017815.1 XP_016873304.1 8 1012
          XM_017017816.1 XP_016873305.1 6 782
          XM_017017817.1 XP_016873306.1 6 707
          XM_017017818.1 XP_016873307.1 4 358
          XM_017017819.1 XP_016873308.1 4 303
          XM_017017820.1 XP_016873309.1 7 748
          XM_017017821.1 XP_016873310.1 7 744
          XM_017017822.1 XP_016873311.1 5 516
          XM_017017823.1 XP_016873312.1 6 620
          XM_017017824.1 XP_016873313.1 6 618
          XM_017017825.1 XP_016873314.1 5 536
          XM_017017826.1 XP_016873315.1 5 549
          XM_017017827.1 XP_016873316.1 7 975
          XM_017017828.1 XP_016873317.1 6 605
          XM_017017829.1 XP_016873318.1 4 381
          GenBank mRNA AB231311.1 - 3 268
          AF052642.1 - 3 268
          AF052643.1 - 3 268
          AF052644.1 - 3 268
          AF450266.1 - 5 482
          AK093209.1 - 11 1509
          AK128739.1 - 9 2180
          AK291507.1 - 5 543
          DA816251.1 - 5 543
          FJ224327.1 - 5 487
          X76732.1 - 5 487

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          calmodulin-related

          Gene Ontology Categories:

          Process(es)

          DNA binding
          calcium ion binding
          protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-qfdf2:80/100.66.76.55:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline