Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other NKX3-1 SIRNA Assays ›
          Gene Symbol:
          Nkx3-1
          Gene Name:
          NK-3 transcription factor, locus 1 (Drosophila)
          Gene Aliases:
          Bax, NKX3.1, NKX3A, Nkx-3.1, bagpipe
          Chromosome Location:
          Chr.14: 69190692 - 69194658 on Build GRCm38
          UniGene ID:
          Mm.3520
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 156537

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Nkx3-1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010921.3 NP_035051.1 2 564
          GenBank mRNA AK137281.1 - 2 575
          BC050109.1 - 2 561
          BC132401.1 - 2 618 40130678
          BC132427.1 - 2 618 40130704
          U73460.1 - 2 551
          U88542.1 - 2 564

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          homeodomain transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          urogenital system development
          positive regulation of protein phosphorylation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          multicellular organism development
          salivary gland development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          positive regulation of gene expression
          negative regulation of gene expression
          positive regulation of cell death
          positive regulation of phosphatidylinositol 3-kinase signaling
          androgen receptor signaling pathway
          prostate gland development
          response to testosterone
          internal genitalia morphogenesis
          positive regulation of cysteine-type endopeptidase activity involved in apoptotic process
          steroid hormone mediated signaling pathway
          protein kinase B signaling
          negative regulation of insulin-like growth factor receptor signaling pathway
          positive regulation of protein kinase activity
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          negative regulation of mitotic cell cycle
          positive regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          branching morphogenesis of an epithelial tube
          negative regulation of epithelial cell proliferation
          positive regulation of cell division
          branching involved in prostate gland morphogenesis
          epithelial cell proliferation involved in salivary gland morphogenesis
          negative regulation of epithelial cell proliferation involved in prostate gland development
          cellular response to steroid hormone stimulus
          cellular response to hypoxia
          mitotic cell cycle arrest
          negative regulation of estrogen receptor binding
          positive regulation of androgen secretion
          positive regulation of response to DNA damage stimulus
          positive regulation of apoptotic signaling pathway
          positive regulation of intrinsic apoptotic signaling pathway
          transcription regulatory region sequence-specific DNA binding
          transcription factor activity, RNA polymerase II core promoter sequence-specific
          core promoter binding
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          androgen receptor activity
          protein binding
          transcription factor binding
          cysteine-type endopeptidase activator activity involved in apoptotic process
          estrogen receptor activity
          protein kinase activator activity
          estrogen receptor binding
          histone deacetylase binding
          sequence-specific DNA binding
          protein self-association
          transcription regulatory region DNA binding
          MADS box domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-t5qbq:80/100.66.79.226:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0