Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other ABCG1 SIRNA Assays ›
          Gene Symbol:
          ABCG1
          Gene Name:
          ATP binding cassette subfamily G member 1
          Gene Aliases:
          ABC8, WHITE1
          Chromosome Location:
          Chr.21: 42199689 - 42304389 on Build GRCh38
          UniGene ID:
          Hs.124649
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 15658

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: ABCG1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_004915.3 NP_004906.3 15 2635
          NM_016818.2 NP_058198.2 15 2599
          NM_207174.1 NP_997057.1 15 2600
          NM_207627.1 NP_997510.1 16 2759
          NM_207628.1 NP_997511.1 17 2677
          NM_207629.1 NP_997512.1 15 2563
          XM_011529806.1 XP_011528108.1 15 2713
          XM_011529807.2 XP_011528109.1 15 2735
          GenBank mRNA AF038175.1 - 5 1193
          AF323660.1 - 16 2762
          AF323661.1 - 17 2680
          AF323662.1 - 15 2566
          AF323663.1 - 15 2525
          AF323664.1 - 15 2603
          AK092959.1 - 11 2475
          AL833689.1 - 1 1314
          AY048757.1 - 16 3118
          BC029158.1 - 15 2600 5198758
          X91249.1 - 15 2550

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          ATP-binding cassette (ABC) transporter primary active transporter transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          regulation of transcription, DNA-templated
          cholesterol metabolic process
          detection of hormone stimulus
          response to organic substance
          negative regulation of macrophage derived foam cell differentiation
          regulation of cholesterol esterification
          positive regulation of cholesterol efflux
          negative regulation of cholesterol storage
          intracellular cholesterol transport
          cholesterol efflux
          phospholipid efflux
          response to lipid
          low-density lipoprotein particle remodeling
          high-density lipoprotein particle remodeling
          glycoprotein transport
          lipoprotein metabolic process
          cholesterol homeostasis
          amyloid precursor protein catabolic process
          reverse cholesterol transport
          positive regulation of cholesterol biosynthetic process
          transmembrane transport
          phospholipid homeostasis
          response to high density lipoprotein particle
          toxin transport
          ATP binding
          phospholipid binding
          phospholipid transporter activity
          cholesterol binding
          cholesterol transporter activity
          toxin transporter activity
          sterol-transporting ATPase activity
          glycoprotein transporter activity
          protein homodimerization activity
          ADP binding
          protein heterodimerization activity
          protein dimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0