Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CTNS SIRNA Assays ›
          Gene Symbol:
          CTNS
          Gene Name:
          cystinosin, lysosomal cystine transporter
          Gene Aliases:
          CTNS-LSB, PQLC4
          Chromosome Location:
          Chr.17: 3636391 - 3663103 on Build GRCh38
          UniGene ID:
          Hs.187667
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 15671

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CTNS
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001031681.2 NP_001026851.2 10, 11 1330
          NM_004937.2 NP_004928.2 10, 11 1453
          XM_005256485.2 XP_005256542.1 10, 11 1453
          XM_006721463.2 XP_006721526.1 10, 11 1402
          XM_006721464.2 XP_006721527.1 9, 10 1368
          XM_011523691.2 XP_011521993.1 10, 11 1252
          XM_011523692.2 XP_011521994.1 9, 10 1373
          XM_017024254.1 XP_016879743.1 8, 9 1289
          XM_017024255.1 XP_016879744.1 9, 10 1368
          XM_017024256.1 XP_016879745.1 9, 10 1373
          XM_017024257.1 XP_016879746.1 8, 9 1289
          XM_017024258.1 XP_016879747.1 8, 9 1288
          GenBank mRNA AJ222967.1 - 10, 11 1199
          AK292019.1 - 10, 11 1166
          AK296178.1 - 7, 8 924
          AK297525.1 - 7, 8 946
          BC032850.2 - 10, 11 1319 5268683
          BX369529.1 - 3, 4 399
          CR542058.1 - 8, 9 860
          KU178003.1 - 8, 9 860
          KU178004.1 - 8, 9 860
          KU178005.1 - 8, 9 842
          KU178006.1 - 7, 8 775

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001191647.

          Panther Classification:

          Molecular Function -

          amino acid transporter transporter secondary carrier transporter

          Gene Ontology Categories:

          Function(s) Process(es)

          lens development in camera-type eye
          cellular amino acid metabolic process
          glutathione metabolic process
          ion transport
          brain development
          long-term memory
          grooming behavior
          adult walking behavior
          visual learning
          L-cystine transport
          melanin biosynthetic process
          ATP metabolic process
          cognition
          transmembrane transport
          L-cystine transmembrane transporter activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0