Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PAFAH1B1 SIRNA Assays ›
          Gene Symbol:
          Pafah1b1
          Gene Name:
          platelet-activating factor acetylhydrolase, isoform 1b, subunit 1
          Gene Aliases:
          LIS-1, Lis1, MMS10-U, Mdsh, Ms10u, Pafaha
          Chromosome Location:
          Chr.11: 74673949 - 74724384 on Build GRCm38
          UniGene ID:
          Mm.397111
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 156886

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pafah1b1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_013625.4 NP_038653.1 11 2467
          GenBank mRNA AK147207.1 - 11 2405
          AK147229.1 - 9 5518
          AK147237.1 - 11 2460
          AK147309.1 - 10 4117
          AK147507.1 - 11 2741
          AK147515.1 - 11 2455
          AK147533.1 - 11 2434
          AK147793.1 - 11 2129
          AK152284.1 - 1 508
          AK161751.1 - 1 102
          AK164276.1 - 1 585
          AK168929.1 - 11 2107
          L25108.1 - 11 2367
          U95116.1 - 11 2762

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          establishment of mitotic spindle orientation
          microtubule cytoskeleton organization
          ameboidal-type cell migration
          acrosome assembly
          neuron migration
          positive regulation of cytokine-mediated signaling pathway
          lipid metabolic process
          transport
          microtubule-based process
          cell cycle
          mitotic nuclear division
          nuclear migration
          chemical synaptic transmission
          multicellular organism development
          nervous system development
          neuroblast proliferation
          learning or memory
          retrograde axonal transport
          adult locomotory behavior
          protein secretion
          negative regulation of neuron projection development
          lipid catabolic process
          cell migration
          stem cell division
          transmission of nerve impulse
          corpus callosum morphogenesis
          hippocampus development
          layer formation in cerebral cortex
          cerebral cortex neuron differentiation
          cerebral cortex development
          actin cytoskeleton organization
          cell differentiation
          microtubule organizing center organization
          osteoclast development
          regulation of GTPase activity
          positive regulation of axon extension
          positive regulation of mitotic cell cycle
          negative regulation of JNK cascade
          vesicle transport along microtubule
          brain morphogenesis
          neuromuscular process controlling balance
          nuclear envelope disassembly
          cell division
          establishment of centrosome localization
          protein binding
          microtubule binding
          protein complex binding
          protein homodimerization activity
          dynein binding
          dynein intermediate chain binding
          phosphoprotein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0