Search
Search

Ly75| Gene Symbol: Ly75 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013825.3 | NP_038853.2 | 4 | 660 | ||
| GenBank mRNA | AF395445.1 | - | 4 | 660 | ||
| AK049301.1 | - | 4 | 703 | |||
| AK144922.1 | - | 4 | 713 | |||
| AK157921.1 | - | 4 | 724 | |||
| BC150734.1 | - | 681 | 9087645 | |||
| U19271.1 | - | 694 | ||||