Search
Search

Lsp1| Gene Symbol: Lsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136071.2 | NP_001129543.1 | 6 | 686 | ||
| NM_001271508.1 | NP_001258437.1 | 6 | 1035 | |||
| NM_001271509.1 | NP_001258438.1 | 6 | 668 | |||
| NM_001271510.1 | NP_001258439.1 | 6 | 686 | |||
| NM_001271523.1 | NP_001258452.1 | 6 | 867 | |||
| NM_019391.3 | NP_062264.1 | 6 | 1053 | |||
| GenBank mRNA | AK003081.1 | - | 6 | 616 | ||
| AK075783.1 | - | 6 | 868 | |||
| BC003796.1 | - | 6 | 727 | 3488528 | ||
| BC044736.1 | - | 6 | 641 | |||
| D49691.1 | - | 6 | 690 | |||
| M89956.1 | - | 6 | 628 | |||
| M90316.1 | - | 6 | 1052 | |||
| S74179.1 | - | 6 | 599 | |||