Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other GRN SIRNA Assays ›
          Gene Symbol:
          Grn
          Gene Name:
          granulin
          Gene Aliases:
          Pgrn, epithelin
          Chromosome Location:
          Chr.11: 102430322 - 102436809 on Build GRCm38
          UniGene ID:
          Mm.1568
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 157910

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Grn
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008175.4 NP_032201.2 9 1094
          GenBank mRNA AK013678.1 - 9 910
          AK018744.1 - 9 935
          AK147932.1 - 9 911
          AK149570.1 - 9 913
          AK150125.1 - 9 909
          AK150205.1 - 9 910
          AK150338.1 - 9 899
          AK150420.1 - 9 898
          AK150717.1 - 9 910
          AK150746.1 - 9 911
          AK150881.1 - 9 911
          AK151007.1 - 9 910
          AK151162.1 - 9 911
          AK151343.1 - 9 910
          AK151713.1 - 9 910
          AK151756.1 - 9 910
          AK152048.1 - 9 910
          AK152087.1 - 9 910
          AK152118.1 - 9 899
          AK152213.1 - 9 899
          AK152705.1 - 9 910
          AK152765.1 - 9 910
          AK152944.1 - 9 899
          AK153183.1 - 9 899
          AK153269.1 - 9 910
          AK153289.1 - 9 910
          AK153418.1 - 9 910
          AK153470.1 - 9 910
          AK154492.1 - 9 910
          AK159397.1 - 9 899
          AK159428.1 - 9 899
          AK159557.1 - 9 910
          AK159809.1 - 9 910
          AK160022.1 - 9 899
          AK160033.1 - 9 910
          AK160074.1 - 9 899
          AK182648.1 - 395
          AK211032.1 - 4 363
          AK219696.1 - 4 373
          BC037047.1 - 8 891
          BC049943.1 - 9 896
          BC129849.1 - 9 894 40109630
          BC129850.1 - 9 894 40109631
          M86736.1 - 9 892
          X62321.1 - 8 875

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          blastocyst hatching
          embryo implantation
          positive regulation of neuron projection development
          response to estradiol
          chondrocyte proliferation
          synaptic vesicle endocytosis
          positive regulation of epithelial cell proliferation
          male mating behavior
          positive regulation of dendritic spine development
          neural precursor cell proliferation
          cytokine activity
          poly(A) RNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-pnd5g:80/100.66.79.83:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline