Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC1 SIRNA Assays ›
          Gene Symbol:
          Hdac1
          Gene Name:
          histone deacetylase 1
          Gene Aliases:
          HD1, Hdac1-ps, MommeD5, RPD3
          Chromosome Location:
          Chr.4: 129516104 - 129542646 on Build GRCm38
          UniGene ID:
          Mm.202504
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 158011

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hdac1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008228.2 NP_032254.1 5 394
          GenBank mRNA AK076080.1 - 5 379
          BC092070.1 - 5 389 4217199
          BC092285.1 - 5 394
          BC108371.1 - 5 395 4976514
          U80780.1 - 5 391
          X98207.1 - 5 394

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          response to amphetamine
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          protein deacetylation
          endoderm development
          circadian rhythm
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          epidermal cell differentiation
          negative regulation of gene expression
          positive regulation of receptor biosynthetic process
          chromatin modification
          histone deacetylation
          hippocampus development
          neuron differentiation
          positive regulation of interleukin-1 production
          positive regulation of tumor necrosis factor production
          circadian regulation of gene expression
          cellular response to oxidative stress
          odontogenesis of dentin-containing tooth
          positive regulation of tyrosine phosphorylation of Stat3 protein
          embryonic digit morphogenesis
          ATP-dependent chromatin remodeling
          negative regulation of apoptotic process
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of neuron apoptotic process
          negative regulation by host of viral transcription
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of insulin secretion
          rhythmic process
          positive regulation of oligodendrocyte differentiation
          negative regulation of androgen receptor signaling pathway
          hair follicle placode formation
          eyelid development in camera-type eye
          fungiform papilla formation
          histone H3 deacetylation
          histone H4 deacetylation
          negative regulation of canonical Wnt signaling pathway
          positive regulation of chemokine (C-X-C motif) ligand 2 production
          positive regulation of type B pancreatic cell apoptotic process
          negative regulation of peptidyl-lysine acetylation
          negative regulation of intrinsic apoptotic signaling pathway
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II core promoter proximal region sequence-specific DNA binding
          RNA polymerase II distal enhancer sequence-specific DNA binding
          core promoter binding
          RNA polymerase II repressing transcription factor binding
          RNA polymerase II transcription corepressor activity
          DNA binding
          chromatin binding
          transcription factor activity, sequence-specific DNA binding
          transcription corepressor activity
          histone deacetylase activity
          protein binding
          transcription factor binding
          hydrolase activity
          deacetylase activity
          enzyme binding
          histone deacetylase activity (H3-K14 specific)
          nucleosomal DNA binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          histone deacetylase activity (H3-K9 specific)
          protein complex binding
          protein deacetylase activity
          activating transcription factor binding
          histone deacetylase activity (H4-K16 specific)
          Krueppel-associated box domain binding
          histone deacetylase binding
          transcription regulatory region DNA binding
          NAD-dependent histone deacetylase activity (H3-K9 specific)
          NAD-dependent histone deacetylase activity (H4-K16 specific)
          protein N-terminus binding
          NF-kappaB binding
          repressing transcription factor binding
          NAD-dependent histone deacetylase activity (H3-K18 specific)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0