Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PIK3CD SIRNA Assays ›
          Gene Symbol:
          PIK3CD
          Gene Name:
          phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit delta
          Gene Aliases:
          APDS, IMD14, P110DELTA, PI3K, p110D
          Chromosome Location:
          Chr.1: 9629889 - 9729114 on Build GRCh38
          UniGene ID:
          Hs.518451
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 15812

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: PIK3CD
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_005026.3 NP_005017.3 6 953
          XM_006710687.2 XP_006710750.1 6 1013
          XM_006710689.2 XP_006710752.1 5 867
          XM_017001476.1 XP_016856965.1 6 1059
          XM_017001477.1 XP_016856966.1 7 996
          XM_017001478.1 XP_016856967.1 7 1156
          XM_017001479.1 XP_016856968.1 6 1025
          XM_017001480.1 XP_016856969.1 5 867
          XM_017001481.1 XP_016856970.1 5 940
          XM_017001482.1 XP_016856971.1 5 862
          XM_017001483.1 XP_016856972.1 5 852
          XM_017001484.1 XP_016856973.1 6 1059
          XM_017001485.1 XP_016856974.1 5 852
          GenBank mRNA AB208835.1 - 1 148
          AK314191.1 - 6 954
          BC132919.1 - 5 867 40146941
          BC132921.1 - 5 867 40146943
          BC144264.1 - 5 867 9052784
          BC150297.1 - 6 900 8860364
          DQ263594.1 - 4 745
          DQ263595.1 - 4 745
          DQ263596.1 - 4 745
          JN190435.1 - 5 980
          U57843.1 - 5 860
          U86453.1 - 6 940
          Y10055.2 - 6 941

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          kinase

          Gene Ontology Categories:

          Function(s) Process(es)

          natural killer cell differentiation
          cytokine production
          adaptive immune response
          mast cell chemotaxis
          respiratory burst involved in defense response
          protein phosphorylation
          phosphatidylinositol biosynthetic process
          inflammatory response
          signal transduction
          positive regulation of gene expression
          T cell chemotaxis
          phosphatidylinositol 3-kinase signaling
          regulation of phosphatidylinositol 3-kinase signaling
          natural killer cell activation
          T cell differentiation
          positive regulation of cell migration
          neutrophil chemotaxis
          natural killer cell chemotaxis
          B cell chemotaxis
          phosphatidylinositol-3-phosphate biosynthetic process
          T cell activation
          B cell activation
          mast cell degranulation
          innate immune response
          phosphatidylinositol phosphorylation
          phosphatidylinositol-mediated signaling
          T cell receptor signaling pathway
          B cell receptor signaling pathway
          mast cell differentiation
          neutrophil extravasation
          protein binding
          ATP binding
          1-phosphatidylinositol-3-kinase activity
          phosphatidylinositol 3-kinase activity
          1-phosphatidylinositol-4-phosphate 3-kinase activity
          phosphatidylinositol-4,5-bisphosphate 3-kinase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0