Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TLE1 SIRNA Assays ›
          Gene Symbol:
          TLE1
          Gene Name:
          transducin like enhancer of split 1
          Gene Aliases:
          ESG, ESG1, GRG1
          Chromosome Location:
          Chr.9: 81583683 - 81689535 on Build GRCh38
          UniGene ID:
          Hs.197320
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 15839

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TLE1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001303103.1 NP_001290032.1 5, 6 1597
          NM_001303104.1 NP_001290033.1 5, 6 1597
          NM_005077.4 NP_005068.2 5, 6 1597
          XM_005252151.1 XP_005252208.1 5, 6 511
          XM_005252152.1 XP_005252209.1 5, 6 511
          XM_005252153.1 XP_005252210.1 5, 6 511
          XM_005252154.1 XP_005252211.1 5, 6 511
          XM_005252156.2 XP_005252213.1 5, 6 511
          XM_005252162.1 XP_005252219.1 5, 6 511
          XM_005252163.2 XP_005252220.1 5, 6 511
          XM_006717258.1 XP_006717321.1 5, 6 511
          XM_006717260.1 XP_006717323.1 5, 6 511
          XM_006717261.2 XP_006717324.1 3, 4 223
          XM_006717262.1 XP_006717325.1 5, 6 511
          XM_006717263.1 XP_006717326.1 5, 6 511
          XM_011518951.2 XP_011517253.1 5, 6 511
          XM_017015064.1 XP_016870553.1 5, 6 511
          XM_017015065.1 XP_016870554.1 5, 6 511
          GenBank mRNA AB209854.1 - 5, 6 1597
          AK290860.1 - 5, 6 359
          AK293606.1 - 5, 6 385
          AK304565.1 - 5, 6 420
          BC010100.2 - 5, 6 743 3506451
          BC015747.1 - 5, 6 453 4857394
          DB211613.1 - 3, 4 170
          M99435.1 - 5, 6 327

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001173433, XM_008763792, XM_008763793, XM_006238281, XM_008763789, and others.
          This siRNA also targets the following transcripts in Mouse: NM_001285530, NM_011599, NM_001285529, NM_001285531, NM_001285532, and others.

          Panther Classification:

          Molecular Function -

          transcription cofactor

          Gene Ontology Categories:

          Function(s) Process(es)

          negative regulation of transcription from RNA polymerase II promoter
          transcription, DNA-templated
          signal transduction
          multicellular organism development
          organ morphogenesis
          positive regulation of gene expression
          Wnt signaling pathway
          negative regulation of Wnt signaling pathway
          negative regulation of I-kappaB kinase/NF-kappaB signaling
          negative regulation of transcription, DNA-templated
          beta-catenin-TCF complex assembly
          negative regulation of anoikis
          RNA polymerase II transcription corepressor activity
          protein binding
          transcription factor binding
          identical protein binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0