Search
Search

Ext1| Gene Symbol: Ext1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010162.2 | NP_034292.2 | 1, 2 | 1778 | ||
| GenBank mRNA | AK132327.1 | - | 1, 2 | 1755 | ||
| BC004741.1 | - | 1, 2 | 1142 | 3499743 | ||
| U78539.1 | - | 1, 2 | 1282 | |||
| X96639.1 | - | 1, 2 | 1552 | |||