Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FGR SIRNA Assays ›
          Gene Symbol:
          Fgr
          Gene Name:
          Gardner-Rasheed feline sarcoma viral (Fgr) oncogene homolog
          Gene Aliases:
          -
          Chromosome Location:
          Chr.4: 132974095 - 133001893 on Build GRCm38
          UniGene ID:
          Mm.271665
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 158534

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Fgr
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010208.4 NP_034338.3 10 1272
          GenBank mRNA AK036438.1 - 10 1194
          AK036476.1 - 10 1444
          AK038141.1 - 10 1178
          AK089812.1 - 10 1242
          AK155902.1 - 10 1248
          AK157640.1 - 1226
          AK171824.1 - 10 1226
          BC057863.1 - 10 1141 5317449
          X16440.1 - 10 1207
          X52191.1 - 10 1179

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          protein phosphorylation
          transmembrane receptor protein tyrosine kinase signaling pathway
          integrin-mediated signaling pathway
          regulation of cell shape
          positive regulation of phosphatidylinositol 3-kinase signaling
          phosphorylation
          peptidyl-tyrosine phosphorylation
          cell differentiation
          positive regulation of cell migration
          regulation of actin cytoskeleton organization
          peptidyl-tyrosine autophosphorylation
          regulation of cell proliferation
          positive regulation of mast cell degranulation
          positive regulation of phosphatidylinositol 3-kinase activity
          innate immune response
          regulation of innate immune response
          regulation of protein kinase activity
          protein autophosphorylation
          positive regulation of cytokine secretion
          regulation of phagocytosis
          defense response to Gram-positive bacterium
          cellular response to peptide hormone stimulus
          nucleotide binding
          phosphotyrosine binding
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          protein binding
          ATP binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein kinase binding
          immunoglobulin receptor binding
          Fc-gamma receptor I complex binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0