Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HDAC6 SIRNA Assays ›
          Gene Symbol:
          Hdac6
          Gene Name:
          histone deacetylase 6
          Gene Aliases:
          Hd6, Hdac5, Sfc6, mHDA2
          Chromosome Location:
          Chr.X: 7930120 - 7947889 on Build GRCm38
          UniGene ID:
          Mm.29854
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 158921

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hdac6
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130416.1 NP_001123888.1 9, 10 998
          NM_010413.3 NP_034543.3 9, 10 1073
          GenBank mRNA AF006603.2 - 8, 9 758
          AK148092.1 - 9, 10 919
          AK148144.1 - 9, 10 889
          AK154099.1 - 9, 10 937
          AK155545.1 - 9, 10 948
          AK155546.1 - 9, 10 937
          BC041105.1 - 9, 10 921 5352032

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          protein polyubiquitination
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          protein deacetylation
          ubiquitin-dependent protein catabolic process
          misfolded or incompletely synthesized protein catabolic process
          intracellular protein transport
          negative regulation of microtubule depolymerization
          response to toxic substance
          positive regulation of signal transduction
          response to organic substance
          regulation of receptor activity
          positive regulation of epithelial cell migration
          positive regulation of receptor biosynthetic process
          macroautophagy
          chromatin modification
          histone deacetylation
          lysosome localization
          peptidyl-lysine deacetylation
          cellular response to topologically incorrect protein
          regulation of gene expression, epigenetic
          ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
          protein complex disassembly
          negative regulation of protein complex disassembly
          regulation of fat cell differentiation
          negative regulation of proteolysis
          mitochondrion localization
          response to misfolded protein
          regulation of establishment of protein localization
          cellular response to hydrogen peroxide
          aggresome assembly
          polyubiquitinated misfolded protein transport
          Hsp90 deacetylation
          response to growth factor
          cellular response to misfolded protein
          positive regulation of chaperone-mediated protein complex assembly
          tubulin deacetylation
          mitophagy in response to mitochondrial depolarization
          positive regulation of hydrogen peroxide-mediated programmed cell death
          core promoter binding
          actin binding
          histone deacetylase activity
          protein binding
          beta-catenin binding
          microtubule binding
          zinc ion binding
          hydrolase activity
          histone deacetylase activity (H3-K14 specific)
          polyubiquitin binding
          ubiquitin protein ligase binding
          NAD-dependent histone deacetylase activity (H3-K14 specific)
          histone deacetylase activity (H3-K9 specific)
          histone deacetylase activity (H4-K16 specific)
          histone deacetylase binding
          tubulin deacetylase activity
          alpha-tubulin binding
          ubiquitin binding
          metal ion binding
          NAD-dependent histone deacetylase activity (H3-K9 specific)
          NAD-dependent histone deacetylase activity (H4-K16 specific)
          tau protein binding
          beta-tubulin binding
          misfolded protein binding
          Hsp90 protein binding
          dynein complex binding
          NAD-dependent histone deacetylase activity (H3-K18 specific)

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0