Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other HMGB1 SIRNA Assays ›
          Gene Symbol:
          Hmgb1 Gm12568
          Gene Name:
          high mobility group box 1
          high mobility group box 1 pseudogene
          Gene Aliases:
          DEF, HMG-1, Hmg1, SBP-1, amphoterin, p30
          OTTMUSG00000007516
          Chromosome Location:
          Chr.5: 149047222 - 149053037 on Build GRCm38
          Chromosome Location:
          Chr.11: 46243071 - 46244300 on Build GRCm38
          UniGene ID:
          Mm.207047
          Mm.455099
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          -
          siRNA ID 158976

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Hmgb1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_010439.3 NP_034569.1 5 1197
          GenBank mRNA AK034535.1 - 1 346
          AK051800.1 - 5 1124
          AK084841.1 - 5 1199
          AK087949.1 - 5 1160
          AK088347.1 - 1 96
          AK134228.1 - 4 936
          AK144428.1 - 1175
          AK146420.1 - 5 1142
          AK150924.1 - 5 1202
          AK157846.1 - 5 1199
          AK166007.1 - 5 1153
          BC006586.1 - 5 1130 3587109
          BC008565.1 - 5 1168 3586745
          BC064790.1 - 1483 30019980
          BC083067.1 - 5 1125 5372544
          BC085090.1 - 5 1157 6485915
          BC094030.1 - 5 1194 4952464
          BC100312.1 - 5 1178 30849071
          BC110667.1 - 4 1605 5036554
          BC111468.1 - 5 1128 4215684
          L38477.1 - 5 1113
          U00431.1 - 5 1185
          Gene Symbol: Gm12568
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          GenBank mRNA BC118540.1 - 1004

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          eye development
          positive regulation of protein phosphorylation
          positive regulation of mesenchymal cell proliferation
          chromatin remodeling
          lung development
          positive regulation of cell migration
          DNA geometric change
          positive regulation of myeloid cell differentiation
          positive regulation of glycogen catabolic process
          positive regulation of protein kinase activity
          positive regulation of mitotic cell cycle
          positive regulation of transcription from RNA polymerase II promoter
          induction of positive chemotaxis
          response to glucocorticoid
          open form four-way junction DNA binding
          crossed form four-way junction DNA binding
          bent DNA binding
          chromatin binding
          cytokine activity
          calcium-dependent protein kinase regulator activity
          protein kinase activator activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0