Search
Search

Hspa9| Gene Symbol: Hspa9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010481.2 | NP_034611.2 | 17 | 2545 | ||
| GenBank mRNA | AK004946.1 | - | 17 | 2507 | ||
| AK030299.1 | - | 17 | 2539 | |||
| AK145965.1 | - | 17 | 2539 | |||
| AK159791.1 | - | 17 | 2547 | |||
| AK161587.1 | - | 17 | 2548 | |||
| AK165958.1 | - | 17 | 2545 | |||
| AK167856.1 | - | 17 | 2538 | |||
| BC052727.1 | - | 17 | 2545 | 6813991 | ||
| BC057343.1 | - | 17 | 2545 | 6813991 | ||
| D17556.1 | - | 17 | 2517 | |||