Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INPP5D SIRNA Assays ›
          Gene Symbol:
          Inpp5d
          Gene Name:
          inositol polyphosphate-5-phosphatase D
          Gene Aliases:
          SHIP, SHIP-1, SHIP1, SIP-145, p150Ship, s-SHIP
          Chromosome Location:
          Chr.1: 87620312 - 87720510 on Build GRCm38
          UniGene ID:
          Mm.15105
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 159182

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Inpp5d
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001110192.2 NP_001103662.1 21, 22 2549
          NM_001110193.2 NP_001103663.1 21, 22 2552
          NM_010566.3 NP_034696.2 21, 22 2552
          GenBank mRNA AF125996.1 - 21, 22 2465
          AF184912.1 - 16, 17 1745
          AF184913.1 - 16, 17 1745
          AF228679.1 - 21, 22 2463
          AK143560.1 - 21, 22 2489
          BC108328.1 - 21, 22 2425 30839591
          U39203.1 - 21, 22 2501
          U51742.1 - 21, 22 2463
          U52044.1 - 21, 22 2485

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_019311, XM_008767293, U55192.

          Gene Ontology Categories:

          Function(s) Process(es)

          immune system process
          apoptotic process
          negative regulation of cell proliferation
          determination of adult lifespan
          negative regulation of signal transduction
          immunoglobulin mediated immune response
          dephosphorylation
          negative regulation of granulocyte differentiation
          negative regulation of B cell proliferation
          intracellular signal transduction
          positive regulation of apoptotic process
          negative regulation of interleukin-6 biosynthetic process
          positive regulation of B cell differentiation
          positive regulation of lymphocyte differentiation
          positive regulation of erythrocyte differentiation
          negative regulation of monocyte differentiation
          negative regulation of neutrophil differentiation
          negative regulation of osteoclast differentiation
          negative regulation of bone resorption
          phosphatidylinositol dephosphorylation
          negative regulation of immune response
          negative regulation of B cell activation
          inositol-polyphosphate 5-phosphatase activity
          protein binding
          hydrolase activity
          SH3 domain binding
          inositol-4,5-bisphosphate 5-phosphatase activity
          phosphatidylinositol trisphosphate phosphatase activity
          PTB domain binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0